Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Product Code. 11871020
Click to view available options
Range:
0 to 500 mg/L Total
0.00 to 5.00 mg/L Total
0.000 to 0.500 mg/L Free
No. of Tests:
100
300
600
For Use With (Application):
Chlorine Ultra High Range
Free Chlorine Ultra Low Range
Total Chlorine
Total Chlorine (DPD3)
Total Chlorine Ultra Low Range
Unit Size:
Each
Pack of 100
Product Code. 11871020
Brand: Hanna Instruments™ HI9576201
Only null left
Add to basket
Add to basket
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
  • Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
For Use With (Application) Free Chlorine Ultra Low Range
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes 100 individual packets of HI95762-0
No. of Tests 100
Quantity 1 Set
Range 0.000 to 0.500 mg/L Free
Test Method Standard Method 4500-Cl G
Type Water Testing Kit