missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing

Product Code. 11612620
Click to view available options
Range:
0 to 500 mg/L Total
0.00 to 5.00 mg/L Total
0.000 to 0.500 mg/L Free
No. of Tests:
100
300
600
For Use With (Application):
Chlorine Ultra High Range
Free Chlorine Ultra Low Range
Total Chlorine
Total Chlorine (DPD3)
Total Chlorine Ultra Low Range
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 11612620

Brand: Hanna Instruments™ HI93711D3

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.

These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.

  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
  • Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)

Specifications

For Use With (Application) Total Chlorine (DPD3)
For Use With (Equipment) Compatible Hanna benchtop and portable photometers (HI83200, HI83203, HI83205, HI83206, HI83208, HI83099, HI96724)
Includes Dropper bottles of HI93701A-T(F), HI93701B-T(F) and HI93711-D3
No. of Tests 600
Quantity 1 Set
Range 0.00 to 5.00 mg/L Total
Test Method USEPA method 330.5 and Standard Method 4500-Cl G
Type Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • DPD3 REAGENTS KIT FOR 600 TESTS (T
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Potassium iodide, Ethylenediaminetetraacetic acid disodium salt dihydrate, N,N-Diethyl-1,4-phenylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.