Learn More
Novus Biologicals™ INSM1 Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-08139
Product Code. 18076449
  • 766.72€ / 0.10mg
Explore more special offers
Description
Description
The INSM1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_2187. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens INSM1, mRNA.
Native