Learn More
Mettler Toledo™ InPro™ 3253i/SG pH Electrodes
Low-maintenance pH electrode supplied complete with factory-filled, prepressurized liquid electrolyte. Mettler Toledo™ InPro™ 3253i/SG pH Electrodes are suited to a wide scope of applications in biotechnology, pharmaceutical and chemical process industries.
1420.00€
Specifications
Connector Types K8S
Diameter (Metric) 12 mm
Type InPro™ 3253i/SG pH Electrode
Includes Solution Ground
Material Glass
View More Specs
Products 1
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
12869031
View Documents
Mettler Toledo™
52005377
120 mm
1420.00€
Each
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
Description
Description
  • Extended operating life
  • Precise measurement values
  • Resistance to measurement problems at the diaphragm
  • Built-in temperature sensor
Specifications
Specifications
K8S
InPro™ 3253i/SG pH Electrode
Glass
0°C to 100°C
12 mm
Solution Ground
0 to 14
CE Certified