Learn More
WTW™ inoLab™ Multi 9620 IDS Multi-Parameter Benchtop Meter
GREENER_CHOICE
Use this benchtop meter for simultaneous measurement with two independent measuring channels. The WTW™ inoLab™ Multi 9620 IDS Multi-Parameter Benchtop Meter is wireless-ready and offers fatigue-proof reading on a brilliant colour display.
Brand:  WTW™ 1FD560
Product Code. 15552016
  • 2390.00€ / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Digital high performance multi-parameter device with two inputs for pH, ISE, conductivity and dissolved oxygen and comprehensive GLP functions
  • Modern high-performance multi-parameter meter with two galvanically isolated measuring channels pH/mV, ISE (including incremental methods), conductivity and oxygen measurement
  • With a large 16:9 colour graphic display under a hygienic glass shield and antibacterial keyboard for use, even in critical areas
  • Use the new wireless modules together with the new IDS plug head sensors to be independent from cables and measure with ease under laboratory hoods or laminar flow benches
  • The massive zinc die casting lower housing adds safety against interference and ensure a safe, stable stand on the lab bench
  • Two USB interfaces to communicate with the computer, printer or USB memory stick allow the transfer of all data
  • An activatable user administration expands the automatic GLP documentation by additional safety and traceability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2
Meter unit with universal power supply, stand and operating manual, software and UUSB cable
Multi 9620 IDS
Multi-Parameter Benchtop Meter
LCD
Manual 500/10000 automatic
Universal power supply 100 to 240 V, 50/60 Hz
All except ORP