missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Pure pH Electrodes

A glass pH electrode designed for ion-weak-samples such as ultrapure water and deionized water.

Brand:  METTLER TOLEDO™ 30248112

 View more versions of this product

Product Code. 15559601

  • 386.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Pure water
  • Ultrapure water
  • Deionized water
  • Waste water
  • Highly saline solution, sea water
  • Low-ion strength

Accessories & Care

  • Immovable glass sleeve junction
  • LoT membrane glass
  • InLab Pure: refillable FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Pure Pro-ISM: refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Recommended Storage

InLab storage solution

TRUSTED_SUSTAINABILITY
Specifications

Specifications

S7
Glass
Combination Electrode
S7 to BNC (30281915)
LoT Glass
InLab storage solution
120 mm
Immovable Glass Sleeve
1 to 11
Measuring samples at low temperatures or with low ion concentrations
ARGENTHAL with Ag+ trap
InLab™ Pure
0°C to 80°C
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.