missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H7N9 HA (A/Anhui/1/2013) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA581736

Product Code. 16328724

  • 285.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin/HA.

Influenza A H7N9 HA (A/Anhui/1/2013) Polyclonal antibody specifically detects Influenza A H7N9 HA (A/Anhui/1/2013) in Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Influenza A H7N9 HA (A/Anhui/1/2013)
Polyclonal
Unconjugated
flu hemagglutinin; influenza hemagglutinin
Recombinant Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin/HA Protein
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.