Learn More
QIAGEN Indicating FTA™ Micro Card
Click to view available options
For Use With (Application):
For ensics, Transgenic Identification, Transfusion Medicine/HLA Typing
Unit Size:
Pack of 100
Description
- Improves process efficiency by enabling direct amplification for STR analysis due to immediate DNA capture on the FTA card.
- Includes an inert dye, which turns to white when a buccal sample or saliva is added for easy confirmation of sample addition.
- Immediate stabilization and protection of nucleic acids for long term archiving of samples at room temperature.
- Different card options to support manual or automated processing for versatility and incorporation into current workflows.
Specifications
Specifications
| Format | Card |
| Sample Type | DNA |
| For Use With (Application) | For ensics, Transgenic Identification, Transfusion Medicine/HLA Typing |
| Label or Dye | Inhert |
| No. of Samples | 1 |
| Product Type | Indicating FTA Micro Card |
| Quantity | 125 μL |
| Sample Volume | 125μL |
Safety and Handling
Signal Word
- Warning