missing translation for 'onlineSavingsMsg'
Learn More

Leica Microsystems Immersion Oil for Microscopes

Use Leica™ Immersion Oil with microscope to increase the resolving power and aperture of the objective lens.

Brand:  Leica Microsystems 13614800


 View more versions of this product

Product Code. 11964539

  • 65.35€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • 10mL bottle
  • For Leica Microscopes
Specifications

Specifications

Immersion Oil
10 mL
10mL
1 Pack
Type A
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Immersion Oil

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P391-Collect spillage.

Supplemental information
  • MIXTURE LIST-Contains : Butene, homopolymer,Terphenyl, Paraffins(petroleum), normalC5-20, Terphenyl, hydrogenated

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.