missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ExoProStar™ 1-Step

Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. Cytiva illustra ExoProStar™ 1-Step comes in one tube for maximum convenience.

Brand:  Cytiva US77701

Product Code. 11911411

  • 38.08€ / 20 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The ExoProStar 1-Step PCR and Sequence Reaction Clean-Up reagent provides a simple, single-step method for removing unincorporated primers and nucleotides prior to downstream analysis. ExoProStar1-Step combines Illustra Exonuclease I and Alkaline Phosphatase enzymes into a single, stable mix requiring just one pipetting step to set up the clean-up reaction. 
  • One tube with one mix of enzymes delivering PCR and sequence reaction clean-up
  • Just one pipetting step to set up your reaction
  • Fast 30-minute, hands-free protocol
  • 100% recovery of PCR product
  • Scalable for different reaction sizes
  • Automation friendly
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C
Tube
20
Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. For maximum convenience comes in a single tube.
Enzymatic PCR and Sequence reaction cleanup
illustra ExoProStar 1-Step
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions