missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-II/IGF2 Antibody (8H1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
196.00€ - 603.00€
Specifications
| Antigen | IGF-II |
|---|---|
| Clone | 8H1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18760284
|
Novus Biologicals
NBP2-37462 |
0.1 mL |
603.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692466
|
Novus Biologicals
NBP2-37462-0.025ml |
0.025 mL |
196.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| IGF-II | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A | |
| IGF2 | |
| IgG1 | |
| Affinity Purified |
| 8H1 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Mouse | |
| Cell Cycle and Replication, Cytokine Research | |
| PBS with 0.05% Sodium Azide | |
| 3481 | |
| Purified recombinant fragment of human IGF2 (AA: 25-180) expressed in E. Coli. | |
| Primary | |
| 20.1 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title