missing translation for 'onlineSavingsMsg'
Learn More

IDE 1, Tocris Bioscience™

Product Code. 18016032
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Induces definitive endoderm formation in mouse and human ESCs

Induces definitive endoderm formation in mouse and human embryonic stem cells (ESCs) (EC50 = 125 nM). Thought to activate the TGF-β signaling pathway; induces Smad2 phosphorylation and increases levels of Nodal expression.

This compound is mixture of E/Z isomers.

Chemical Identifiers

CAS 1160927-48-9
Molecular Formula C15H18N2O5
Molecular Weight (g/mol) 306.318
InChI Key ABKJCDILEUEJSH-UHFFFAOYSA-N
Synonym ide 1
PubChem CID 66974738
IUPAC Name 2-[(6-carboxyhexanoylhydrazinylidene)methyl]benzoic acid
SMILES C1=CC=C(C(=C1)C=NNC(=O)CCCCCC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 306.31
Percent Purity >98%
Chemical Name or Material IDE 1
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'