missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Stock Tuning Solution, Honeywell Fluka™

Certified Reference Material, 10 mg/L each of Li, Y, Ce, Co, Tl in HNO3 2%

Brand:  Honeywell Fluka E2008-100ML

Product Code. 16654194

This item is not returnable. View return policy

Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ICP-MS Stock Tuning Solution
Certified Reference Materials
Standards for ICP and ICP-MS
<2
Liquid
100 mL
ICP-OES and ICP-MS
1.06 g/cm3
Testing Food, Environmental, Pharma,Biotech, Mining, Mineralogy, Petrochem Industries
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions