Learn More
Liebherr™ Water Filtration Cartridges for Ice Machines
Remove water hardness impurities prior to making ice flakes. Liebherr™ Water Filtration Cartridges for Ice Machines works as a de-scaler, de-carbonizer and de-ironizer, prevention of algae and corrosion, deodorizer and water filter.
86.85€
Specifications
Product Type Anticalculus Flocculator Double Cartridge
For Use With (Equipment) Ice machines
View More Specs
Products 1
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
12130142
View Documents
Liebherr™
KIT2CART
KIT2CART
86.85€
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
Description
Description
Liebherr™ Ice Makers create granular wet ice flakes for rapid cooling. Units with air or water condensers are available (water condensers are recommended for hot environments).
  • The CACF1 water filtration device is essential for removing impurities from the water
  • The complete device includes two in-line holders with 1 cartridge each
    • Flocculator cartridge: transforms calcium carbonates into stable, suspended calcium salts that cannot clump together
    • Filter cartridge: sifts the salts and retains impurities suspended in the water
  • The treatment capacity of the CACF1 unit varies, depending on the hardness of the water
  • Replace the cartridges at least 1x/year or when the pressure between in the inlet and the outlet of the unit drops by ≥500g
  • Replacement cartridges are available separately (mfr. no. KIT2CART)
Specifications
Specifications
Anticalculus Flocculator Double Cartridge
Ice machines