missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-PFI 2 hydrochloride, Tocris Bioscience™
Click to view available options
10mg
Description
Potent and selective SETD7 histone lysine methyltransferase inhibitor (IC50 = 2 nM). Exhibits >1000-fold selectivity over DNMT1 and a panel of 18 other methyltransferases. Inhibits YAP nuclear translocation and function following activation of the Hippo signaling pathway in MCF7 cells.
Negative Control also available.
Specifications
Specifications
Molecular Formula | C23H25F4N3O3S·HCl |
Quantity | 10 mg |
Synonym | (1R)-1-[[3-(Trifluoromethyl)phenyl]methyl]-2-oxo-2-(1-pyrrolidinyl)ethyl]1,2,3,4-tetrahydro-6-isoquinolinesulfonamide hydrochloride |
Formula Weight | 535.98 |
Chemical Name or Material | 8-Fluoro-1,2,3,4-tetrahydro-N-[(1R)-2-oxo-2-(1-pyrrolidinyl)-1-[[3-(trifluoromethyl)phenyl]methyl]ethyl]-6-isoquinolinesulfonamide hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction