missing translation for 'onlineSavingsMsg'
Learn More

Hypophosphorous acid, 50 wt.% solution in wate

Product Code. p-7220487 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
5 L
Unit Size:
1 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 10680839

Brand: Thermo Scientific Acros 201005001

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6303-21-5 | H3O2P | 66.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6303-21-5
Molecular Formula H3O2P
Molecular Weight (g/mol) 66.00
MDL Number MFCD02183592
InChI Key ACVYVLVWPXVTIT-UHFFFAOYSA-N
IUPAC Name phosphinic acid
SMILES O[PH2]=O

Specifications

Melting Point -25.0°C
Boiling Point 108.0°C (1013.0 hPa)
Density 1.2700 g/mL
Packaging Plastic bottle
Specific Gravity 1.2700
Quantity 500 mL
Merck Index 15, 4907
Solubility Information Solubility aq. soln.: soluble. Other solubilities: miscible with ethanol, immiscible with hydrocarbons and halogenated, hydrocarbons
Formula Weight 66.00
Chemical Name or Material Hypophosphorous acid
compliance-icons
Product Identifier
  • Hypophosphorous acid, 50 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.