missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil GOLD™ Preparative Guard Cartridge, 5μm

Provides outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography. Thermo Scientific™ Hypersil GOLD™ HPLC columns offer excellent retention and unique selectivity for aromatic and moderately polar analytes.

Brand:  Thermo Scientific™ 25005-019223

Product Code. 15272218

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3/Pk.
10 mm
Preparative Guard
5 μm
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions