Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 10-200μL
Product Code. 10566375 Shop All Thermo Scientific Products
Click to view available options
Sorbent:
BioBasic C18
BioBasic C4
BioBasic C8
HyperSep HILIC
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep Titanium Dioxide
HyperSep Trypsin
HyperSep Zirconium Dioxide
Unit Size:
Pack of 96
Product Code. 10566375
Brand: Thermo Scientific™ 60109210
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:
Mass spectrometry
Desalting
Protein purification
MALDI
Electrophoresis
HPCE
HPLC
CEC
Tips are available in 1-10µL capacity and 10-200µL capacity.
TRUSTED_SUSTAINABILITY
Specifications
Description BioBasic C8; 10 to 200 uL Volume
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent BioBasic C8
Type SPE Tips
Volume (Metric) 10 to 200 μL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.