Description
- These industrial hand gloves offer Fortix™ Abrasion Resistance Technology for enhanced durability
- Ideal nitrile-coated work gloves with up to 2x the grip for safer handling
- Enhanced breathability for even better comfort
Specifications
Specifications
Coating Material | Nitrile Foam |
Color | Black, Gray |
Cut Resistance Rating | ANSI A1 |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Nylon and Spandex |
Size | 5 |
Style | 11-840 |
Glove Cuff Style | Knit |
Length (Metric) | 200 to 275 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Ansell™ HyFlex 11-840 Gloves >