missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide Solution, 35%, Chem-Lab

Product Code. 11995366
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Designed for laboratory use as an oxidizer, bleaching agent and disinfectant. Hydrogen Peroxide Solution, 35%, Chem-Lab is the simplest peroxide, a strong oxidizer that appears as a clear and slightly viscous liquid. Available in a 5L container.

Hydrogen peroxide’s oxidizing properties are so strong that it is considered a highly reactive oxygen species. Due to it being a weak acid, it can form hydroperoxide or peroxide salts. Industrially, it is used for pulp/paper bleaching, in the manufacture of sodium percarbonate and sodium perborate, and in water purification to remove organic impurities and odors.

Specifications

Color Colorless
CAS 7722-84-1
Boiling Point 110°C
Melting Point -24°C
Molecular Formula H2O2
Composition 35+% H2O2
Density 1.13g/mL
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
Solubility Information Water Soluble
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name hydrogen peroxide
Molecular Weight (g/mol) 34.014
PubChem CID 784
ChEBI CHEBI:16240
Formula Weight 34.01g/mol
Percent Purity >35%
Odor Odorless
pH 2 to 4
Quantity 5 L
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide 35 weight % solution (stab.) p.
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Oxidising liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.