missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, Honeywell™

Product Code. 16626173
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Semiconductor grade (Honeywell 17823), fuming 37%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7647-01-0, 7732-18-5
Boiling Point 42°C (1.013 hPa)
CAS Min % ≥50.00%, ≥30.00%
CAS Max % <70.00%, <50.00%
Packaging Plastic Can
Quantity 2.5 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Beilstein 1098214
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Grade Semiconductor grade acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'