missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.01M, Chem-Lab

Product Code. 10065472
Click to view available options
Quantity:
20 L
Packaging:
EP
This item is not returnable. View return policy
This item is not returnable. View return policy

For laboratory use, Titrations, ASTM D-4047

Specifications

CAS 7647-01-0
Concentration 0.365g HCl / 1L H2O
Density 1.00g/mL
Packaging EP
Normality 0.01N (±0.00002 at 20°C)
Quantity 20 L
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.