Learn More
Chemical Identifiers
CAS | 7335-65-1 |
---|---|
Molecular Formula | C2H8N2O2 |
Molecular Weight (g/mol) | 92.10 |
MDL Number | MFCD00013141 |
InChI Key | YFHNDHXQDJQEEE-UHFFFAOYSA-N |
Synonym | hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph |
PubChem CID | 165591 |
IUPAC Name | acetic acid;hydrazine |
SMILES | NN.CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10125803
|
Thermo Scientific Acros
411750050 |
5 g | Glass bottle |
64.02€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10361301
|
Thermo Scientific Acros
411750250 |
25 g | Glass bottle |
178.89€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
7335-65-1 | |
92.10 | |
YFHNDHXQDJQEEE-UHFFFAOYSA-N | |
165591 | |
NN.CC(O)=O |
C2H8N2O2 | |
MFCD00013141 | |
hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph | |
acetic acid;hydrazine |
Specifications
7335-65-1 | |
Authentic | |
C2H8N2O2 | |
MFCD00013141 | |
YFHNDHXQDJQEEE-UHFFFAOYSA-N | |
acetic acid;hydrazine | |
165591 | |
97% | |
Hydrazine acetate, 99% |
White | |
97% | |
NH2NH2·CH3COOH | |
hydrazine acetate, hydrazinium acetate, hydrazine, monoacetate, hydrazine monoacetate, acetic acid; hydrazine, hydrazineacetate, hydrazine, acetate 1:1, acetic acid; diamine, hydrazine mono-acetate, acmc-209bph | |
NN.CC(O)=O | |
92.10 | |
92.1 | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause cancer.
May cause an allergic skin reaction.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN: Gently wash with plenty of soap and water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 230-845-2
missing translation for 'tsca' : TSCA
- Hydrazine acetate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.