Learn More
Cytiva HyClone™ Tetracycline Screened Characterized Fetal Bovine Serum, US Origin
FBS with undetectable levels of tetracycline
Brand: Cytiva SH30071.03TH
This item is not returnable.
View return policy
Description
Tetracycline Screened FBS is designed for researchers using tetracycline-regulated gene expression systems in cultured cells and is suitable for expression studies in Tet-on/Tet-off systems.Specifications
United States | |
Tetracycline screened | |
Heat Inactivated | |
Yes |
Liquid | |
500 mL | |
≤ 10 EU/mL | |
≤ 10 mg/dL |