Learn More
Cytiva HyClone™ Bovine Calf Serum, US Origin
A cost-effective alternative to Fetal Bovine Serum (FBS)
Brand:  Cytiva SH30072.02NI
 View more versions of this product
Product Code. 16665735
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Collected by venipuncture from formula-fed veal calves
  • Age of animal at time of collection approximately 16-24 weeks
  • Complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Sourced in the USA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
16 to 22 weeks
100 mL
Formula-Fed Veal Calves
≤-10°C
United States
Sequential-filtered, 100 nm
Virus Panel Testing According to 9 CFR 113.53