Learn More
Hamilton™ HVXM Large Body Modular Valve Positioner PTFE Valves
With a larger port internal diameter to allow increased fluid flows.
Brand:  Hamilton™ 36759
 View more versions of this product
Product Code. 11503364
  • 407.00€ / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Description
  • Physically the largest valves
  • Even moderately viscous fluids flow freely
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
Specifications
Specifications
38°C
108.8 μL dead volume
Large body PTFE valve
4-5 model, 4 valves
1/4 in.-28
Aluminum (Housing)