Learn More
R&D Systems™ Human Tyro3/Dtk DuoSet ELISA
Sufficient for at least 15 96-well plates
Brand: R&D Systems™ DY859
This item is not returnable.
View return policy
Description
The Human Tyro3/Dtk DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Dtk/TYRO3.Specifications
Biotin | |
ELISA | |
Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY |
62.50 to 4000 pg/mL | |
7301 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |