Learn More
Invitrogen™ Human TRPC1 (aa 655-787) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP91251
Product Code. 30196523
  • 259.00€ / 100µL
Explore more special offers
Description
Description
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P48995
Blocking Assay, Control
7220
100 μL
RUO
Trpc1
Recombinant
E. coli
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
TRPC1
-20° C, Avoid Freeze/Thaw Cycles
3010009O07Rik; AI115608; capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; membe; Mtrp1; mTrp2; Short transient receptor potential channel 1; smTRPC2; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; transient receptor protein 1; trp; TRP1; TRP-1; TRP-1 protein; trp2; trpc1; trpc1.L; TRPC2a; TRPC2b; trp-related protein 1; Trrp1; Trrp2; XELAEV_18027727mg; xtrpc1
Unconjugated
HEDKEWKFARAKLWLSYFDDKCTLPPPFNIIPSPKTICYMISSLSKWICSHTSKGKVKRQNSLKEWRNLKQKRDENYQKVMCCLVHRYLTSMRQKMQSTDQATVENLNELRQDLSKFRNEIRDLLGFRTSKYA
Suggestions de produits
Suggestions de produits
Vidéos
FDS