Learn More
Novus Biologicals™ Human Stomach Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired)
Brand: Novus Biologicals™ NBL1-26533
This item is not returnable.
View return policy
Description
The Human Stomach Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired) is derived from human.Specifications
Digestive/Waste | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
Human Stomach Whole Tissue Lysate (Adult Primary and Metastatic Tumor Paired) | |
Adult |
1 Set | |
Matched Pair - Total Protein - Human Primary Tumor and Metastatic Tumor Tissue (PM): Stomach | |
Primary and Metastatic Tumor Paired | |
Human | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
For Research Use Only