missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Serpin A5/Protein C Inhibitor ELISA Kit

Human Serpin A5/Protein C Inhibitor quantitates human Serpin A5/Protein C Inhibitor in serum, plasma, supernatant.

656.00€ - 2635.00€

Specifications

Accession Number 4933415L04; Acrosomal serine protease inhibitor; alpha-1 antiproteinase; antitr; antitrypsin; PAI3; PAI-3; Pci; PCI-B; PLANH3; Plasma serine protease inhibitor; Plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; PROCI; Protein C inhibitor; serine (or cysteine) peptidase inhibitor, clade A, member 5; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serine (or cysteine) proteinase inhibitor, clade A, member 5; serpin A5; serpin family A member 5; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serpin peptidase inhibitor, clade A, member 5; Serpina5
Assay Range 24.58 to 6,000 pg/mL
Assay Sensitivity 24 pg/mL
Conjugate Biotin
Product Type ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17105199
View Documents
Invitrogen™
EH412RBX5
5 x 96 Tests
2635.00€
Each
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
 
17174713
View Documents
Invitrogen™
EH412RB
96 Tests
656.00€
Pack of 96
Estimated Shipment: 27-12-2024
to see stock.
Add to basket
 
Description

Description

Human Serpin A5/Protein C Inhibitor quantitates human Serpin A5/Protein C Inhibitor in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin A5/Protein C Inhibitor.

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

4933415L04; Acrosomal serine protease inhibitor; alpha-1 antiproteinase; antitr; antitrypsin; PAI3; PAI-3; Pci; PCI-B; PLANH3; Plasma serine protease inhibitor; Plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; PROCI; Protein C inhibitor; serine (or cysteine) peptidase inhibitor, clade A, member 5; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serine (or cysteine) proteinase inhibitor, clade A, member 5; serpin A5; serpin family A member 5; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serpin peptidase inhibitor, clade A, member 5; Serpina5
24 pg/mL
ELISA
Colorimetric Microplate Reader
SERPINA5
<10%
HRP
Serum, 50 μL; Supernatant, 50 μL; Plasma, 50 μL
SERPINA5
4 hr 45 min
24.58 to 6,000 pg/mL
Biotin
Plasma, Serum, Supernatant
P05154
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Research Use Only
2°C to 8°C
1 hr 20 min
Videos
SDS
Documents

Documents

Product Certifications