Learn More
R&D Systems™ Human SALM1/LRFN2 (NP_065788) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand: R&D Systems™ RDC1579
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human SALM1/LRFN2 (NP_065788) VersaClone cDNA clone contains the complete ORF for LRFN2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LRFN2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature
Specifications
LRFN2 | |
10 μg at 0.2 μg/μL | |
2382bp | |
fibronectin type III, immunoglobulin and leucine rich repeat domains 2, FIGLER2, KIAA1246, leucine rich repeat and fibronectin type III domain containing 2, LRFN2, SALM1 | |
Human |
NP_065788 | |
cDNA is provided in 10mM Tris-Cl, pH 8.252 | |
57497 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
For Reasearch Use Only. Not for use in Humans.