missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RUNX3 (aa 205-221) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP292
This item is not returnable.
View return policy
Description
Human RUNX3 (residues 205-221) Synthetic Peptide, PEP-292, from Invitrogen.
PEP-292 is a 17 amino acid synthetic peptide whose sequence corresponds to residues 205-221 of human RUNX3. The sequence of this peptide is (amino to carboxyl terminus): T (205) K P F P D R F G D L E R L R M (221)R.This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, RUNX3, catalog # PA1-318. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Specifications
864 | |
HPLC | |
Human RUNX3 (aa 205-221) | |
Synthetic Peptide | |
peptide with no preservative | |
Core-binding factor, alpha 3 subunit, CBF-alpha 3, Acute myeloid leukemia 2 protein, Oncogene AML-2, Polyomavirus enhancer-binding protein 2 alpha C | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q13761 | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
RUNX3 | |
No preservative | |
50 μg | |
T(205) K P F P D R F G D L E R L R M (221)R |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction