Learn More
Novus Biologicals™ Human Rectum Tissue MicroArray (Tumor)
The panel contains 5 to 8 different tissues on one slide.
Brand:  Novus Biologicals™ NBP2-78094
Product Code. 18624660
  • 614.00€ / Pack of 5
Explore more special offers
Specifications
5 Slides
Human
Please bake slides at 60C for 30 minutes before use.