Learn More
Novus Biologicals™ Human Rectum Tissue MicroArray (Tumor)
The panel contains 5 to 8 different tissues on one slide.
Brand: Novus Biologicals™ NBP2-78094
This item is not returnable.
View return policy
Specifications
5 Slides | |
Human | |
Please bake slides at 60C for 30 minutes before use. |