Learn More
STEMCELL Technologies Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
For characterization of hPSC-derived progenitor cells of definitive endoderm, hepatic, intestinal, or pancreatic lineages
POA
Specifications
Description Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Pancreatic Cells; Pluripotent Stem Cells
For Use With (Application) Real Time PCR (qPCR)
View More Specs
Products 3
Product Code Brand Format Price Quantity & Availability  
Product Code Brand Format Price Quantity & Availability  
17149291
Encompass_Preferred
View Documents
STEMCELL Technologies
07531
96-well Plate Type 1
POA
1 set
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17159291
Encompass_Preferred
View Documents
STEMCELL Technologies
07533
96-well Plate Type 3 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17169291
Encompass_Preferred
View Documents
STEMCELL Technologies
07534
96-well Plate Type 4
POA
1 set
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • The Human Pluripotent Stem Cell (hPSC)-Derived Endoderm Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of hPSC-derived definitive endodermal progenitor cells and their differentiated progeny including pancreatic, hepatic, and intestinal cells
  • This qPCR array provides the gene expression profile of hPSC-derived endodermal cells following in vitro differentiation, and can aid in development of protocols for directed differentiation towards endodermal lineages
  • Genes were selected based on their demonstrated expression in hPSCs, or in hPSC-derived ectodermal, mesodermal, and endodermal lineage cells (D'Amour et al, including pancreatic (Kroon et al, hepatic (Hay et al, and intestinal progenitor cells (Grapin-Botton & Melton)
Specifications
Specifications
Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Pancreatic Cells; Pluripotent Stem Cells
Real Time PCR (qPCR)