missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human NT-3 Recombinant Protein, PeproTech®

E. coli Recombinant Protein

81.50€ - 5240.00€

Specifications

Accession Number P20783
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 4908
Molecular Weight (g/mol) 27.2
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17843833
View Documents
Gibco™
450-03-100UG
100 μg
1050.00€
100µg
Please to purchase this item. Need a web account? Register with us today!
17894523
View Documents
Gibco™
450-03-10UG
10 μg
198.00€
10µg
Please to purchase this item. Need a web account? Register with us today!
17814543
View Documents
Gibco™
450-03-1MG
1 mg
5240.00€
1mg
Please to purchase this item. Need a web account? Register with us today!
17804393
View Documents
Gibco™
450-03-250UG
250 μg
1836.00€
250µg
Please to purchase this item. Need a web account? Register with us today!
17824623
View Documents
Gibco™
450-03-2UG
2 μg
81.50€
2µg
Please to purchase this item. Need a web account? Register with us today!
17864383
View Documents
Gibco™
450-03-500UG
500 μg
3145.00€
500µg
Please to purchase this item. Need a web account? Register with us today!
17824693
View Documents
Gibco™
450-03-50UG
50 μg
632.00€
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
Specifications

Specifications

P20783
protein with no preservative
27.2
E. coli
RUO
AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3
Human
Recombinant
Lyophilized
ELISA, Functional Assay, Western Blot
4908
Human NT-3
-20°C
<1 EU/μg
Ntf3
Unconjugated
MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT
≥ 98% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.