Learn More
STEMCELL Technologies Human Mesenchymal Stem Cell qPCR Array
For characterization of human MSCs and their differentiation to chondrogenic, adipogenic, or osteogenic lineages
POA - 288.90€
Specifications
Description | Adipocytes; Chondrocytes; Mesenchymal Stem and Progenitor Cells; Osteoblasts |
---|---|
For Use With (Application) | Real Time PCR (qPCR) |
Product Code | Brand | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Price | Quantity & Availability | |||||
17179291
|
STEMCELL Technologies
07541 |
96-well Plate Type 1 |
288.90€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17189291
|
STEMCELL Technologies
07543 |
96-well Plate Type 3 | |||||||
17199291
|
STEMCELL Technologies
07544 |
96-well Plate Type 4 |
POA
1 set |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The Human Mesenchymal Stem Cell Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of human mesenchymal stem and progenitor cells (MSCs) and their differentiated chondrogenic, adipogenic, or osteogenic progeny
- MSCs can be expanded in vitro to generate mesenchymal stromal cell cultures, which, under appropriate conditions, can differentiate into adipocytes, chondrocytes, and osteoblasts
- This qPCR array provides the gene expression profile of MSCs and their trilineage derivatives following in vitro differentiation
Specifications
Adipocytes; Chondrocytes; Mesenchymal Stem and Progenitor Cells; Osteoblasts | |
Real Time PCR (qPCR) |