missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Meprin alpha ELISA Kit
Human Meprin alpha quantitates human Meprin alpha in serum, plasma, supernatant.
656.00€ - 2635.00€
Specifications
Accession Number | AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; Endopeptidase-24.18 subunit alpha; Mep1; MEP-1; Mep1a; Mep-1 A; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; Meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA |
---|---|
Assay Range | 2 to 400 ng/mL |
Assay Sensitivity | 2 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17195089
|
Invitrogen™
EH321RBX5 |
5 x 96 Tests |
2635.00€
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
17104623
|
Invitrogen™
EH321RB |
96 Tests |
656.00€
Pack of 96 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
Description
Human Meprin alpha quantitates human Meprin alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Meprin alpha.
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.Specifications
AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; Endopeptidase-24.18 subunit alpha; Mep1; MEP-1; Mep1a; Mep-1 A; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; Meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA | |
2 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
MEP1A | |
<10% | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
MEP1A | |
4 hr 45 min |
2 to 400 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q16819 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |