missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human LIGHT Recombinant Protein, PeproTech®
Recombinant Protein
Brand: Gibco™ 310-09B-100
This item is not returnable.
View return policy
Description
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.Specifications
O43557 | |
Lyophilized | |
19.3 | |
7 x 15 μg | |
-20°C | |
<1 | |
Recombinant | |
Insect cells |
ELISA, Functional Assay, Western Blot | |
protein with no preservative | |
LIGHT | |
Human LIGHT | |
RUO | |
Unconjugated | |
Determined by its ability to induce IL-8 secretion in human BEAS-2B cells. | |
Purified |