Learn More
Human IL-10 ELISPOT Pair, BD
Convenient set of unlabelled anti-IL-10 capture antibody and a biotinylated anti-IL-10 detection antibody
Brand:  BD Biosciences 551883
Product Code. 15806728
  • 923.00€ / Each
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
Includes: Human IL-10 ELISPOT Capture Antibody (no azide/low endotoxin), Human IL-10 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations