Learn More
Invitrogen™ Human HAL (aa 263-360) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58435 (PA5-58435. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | P42357 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 3034 |
| Name | Human HAL (aa 263-360) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | hal; HIS; histidase; histidine ammonia lyase; histidine ammonia-lyase; histidinemia; Hsd; HSTD; Huth; zgc:136980 |
| Common Name | HAL |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.