Learn More
R&D Systems™ Human GPR180 (NP_851320) VersaClone cDNA
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
Brand:  R&D Systems™ RDC2431
Product Code. 18706899
  • 317.00€ / 10µg
Explore more special offers
Description
Description
  • The Human GPR180 (NP_851320) VersaClone cDNA clone contains the complete ORF for GPR180, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The GPR180 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
Specifications
Specifications
hGPR180
10 μg at 0.2 μg/μL
160897
Human
NP_851320
cDNA is provided in 10 mM Tris-Cl, pH 8.5
Store the unopened product at -20 to -80°C