missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GBAS Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0776
This item is not returnable.
View return policy
Description
NIPSNAP2 Synthetic Peptide, PEP-0776, from Invitrogen.
This peptide corresponds to 14 amino acids near the center of human NIPSNAP2.PEP-0776 can be used as a blocking peptide with polyclonal antibody PA5-20656.Specifications
2631 | |
purified | |
Human GBAS | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O75323 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
NIPSNAP2 | |
0.02% sodium azide | |
50 μg | |
PBS |