Learn More
R&D Systems™ Human FAT10 (NP_006389.2) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand: R&D Systems™ RDC3455
Description
Specifications
FAT10 | |
10537 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_006389.2 | |
Diubiquitin, FAT10diubiquitin, GABBR1, UBD, UBD-3, ubiquitin D, Ubiquitin-like protein FAT10 | |
Human |