Learn More
Invitrogen™ Human Estrogen Receptor alpha (aa 21-32) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP037
Product Code. 11594690
  • 175.00€ / 50µg
Explore more special offers
Description
Description
Human Estrogen Receptor A (ERa) (Residues 21-32) Synthetic Peptide, PEP-037, from Invitrogen.
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. PEP-037 is a 13 amino acid (amino acids. 21-32)synthetic peptide whose sequence corresponds to amino terminal of human estrogen receptor a. The sequence of this peptide is (amino to carboxy terminus): N21-E-L-E-P-L-N-R-P-Q-L-K32- (C) (A cysteine residue was added to facilitate coupling KLH).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human estrogen receptor a, catalog &Num; PA1-309. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2099
purified
Human Estrogen Receptor alpha (aa 21-32)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Human
P03372
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
ESR1
No preservative
50 μg
N21 - E - L - E - P - L - N - R - P - Q - L - K32 - (C)