missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cyclophilin B (aa 194-208) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP058
This item is not returnable.
View return policy
Description
Human Cyclophilin B (residues 194-208) Synthetic Peptide, PEP-058, from Invitrogen.
PEP-058 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 194-208 of human Cyclophilin B. The sequence of this peptide is (amino to carboxy terminus): C194-G-K-I-E-V-E-K-P-F-A-I-A-K-E208This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and Cyclophilin B, catalog # PA1-027A. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
5479 | |
purified | |
Human Cyclophilin B (aa 194-208) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P23284 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
PPIB | |
No preservative | |
50 μg | |
C194 - G - K - I - E - V - E - K - P - F - A - I - A - K - E208 |