missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ATXN7L3 (aa 201-272) Control Fragment Recombinant Protein

Product Code. 30195829
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195829

Brand: Invitrogen™ RP107329

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84638 (PA5-84638. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Component of the transcription regulatory histone acetylation (HAT) complex SAGA, a multiprotein complex that activates transcription by remodeling chromatin and mediating histone acetylation and deubiquitination. Within the SAGA complex, participates in a subcomplex that specifically deubiquitinates both histones H2A and H2B (PubMed:18206972, PubMed:21746879). The SAGA complex is recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation. Within the complex, it is required to recruit USP22 and ENY2 into the SAGA complex (PubMed:18206972). Regulates H2B monoubiquitination (H2Bub1) levels. Affects subcellular distribution of ENY2, USP22 and ATXN7L3B (PubMed:27601583). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14CW9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56970
Name Human ATXN7L3 (aa 201-272) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AT7L3; ataxin 7 like 3; ataxin 7-like 3; ataxin-7-like protein 3; ATXN7L3; DKFZp761G2113; E030022H21Rik; SAGA-associated factor 11 homolog
Common Name ATXN7L3
Gene Symbol ATXN7L3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LRSLLTTQCGVISEHTKKMCTRSLRCPQHTDEQRRTVRIYFLGPSAVLPEVESSLDNDSFDMTDSQALISRL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.