Learn More
Julabo™ HST Booster Heater for Ultra-Low Refrigerated Circulators
3475.00€ - 3665.00€
Specifications
Type | HST Booster Heater |
---|---|
For Use With (Application) | Ideal for controlling the temperature of large or distant objects |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11984798
|
Julabo™
8810011 |
FP51-SL Ultra-Low Refrigerated-Heating CirculatorI |
3475.00€
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
11994798
|
Julabo™
8810012 |
FP52-SL, FP52-SL, FP52-SL-150C, FP55-SL, FP55-SL-150C Ultra-Low Refrigerated-Heating Circulator |
3665.00€
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
Specifications
HST Booster Heater | |
Ideal for controlling the temperature of large or distant objects |