Learn More
Thermo Scientific™ HotStart™ Storage Reaction Tubes
Product Code. p-4526092 Shop All Thermo Scientific Products
Click to view available options
Product Line:
Hot Start Micro 50 Bulk
HotStart Micro 50 Strips
Volume (Metric):
0.2 mL
Unit Size:
Pack of 120
Pack of 480
Product Code. 15292559
Brand: Thermo Scientific™ 6210
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.2 mL
For Use With (Equipment) Thermal Cyclers
Product Line HotStart Micro 50 Strips
Sterility Sterile
Closure Type Flat Cap
Reaction Volume 25 to 50 μL
Product Type Strips