Learn More
Testo™ Hot Wire Probe
Ideal, in conjunction with the appropriate measuring instrument, for determining flow velocity as well as temperature, humidity and pressure in ventilation ducts or at wall or ceiling outlets
Brand:  Testo™ 0635 1543
Product Code. 15808514
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Use the hot wire probe (thermal anemometer) to determine flow velocity and volume flow in ventilation ducts or at wall/ceiling outlets
  • Ø 10 mm
  • 4 functions in 1 probe: measurement of flow, temperature, humidity and absolute pressure
  • With robust, non-rotatable telescope (can be extended to 1100 mm)
  • The immersion depth is easy to read off thanks to the scaling on the telescope
  • For easier handling, the probe can be bent up to an angle of 90°. On the handle there is a practical measuring button, which activates storage of the readings. So you can fully focus on the positioning of the probe in the ventilation duct
  • Intelligent calibration concept
  • The digital probe allows readings to be processed directly in the probe. This technology eliminates instrument measurement uncertainty
  • Flow measuring range: 0 to +20 m/s
Specifications
Specifications
Temperature - ±0.5 °C; Humidity - ±(1.8 %RH + 0.7 % of mv); Pressure - ±3 hPa, Velocity - ±(0.03 m/s + 4 % of mv)
Humidity: 0 to 100%, Pressure - +700 to +1100 hPa; Velocity - 0 to +20 m/s
-20°C to +70°C
1100 mm