missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hot Start Mix RTG
Premixed, predispensed, ambient temperature-stable formulation providing robust and reliable performance for demanding PCR applications in which high specificity and high sensitivity are essential to success
18.35€ - 1039.00€
Specifications
Content And Storage | 1.75 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. Ambient temperature in pouch with desiccant |
---|---|
Format | Tube |
Product Type | illustra Hot Start Mix RTG |
For Use With (Application) | Standard PCR |
Product Code | Brand | Product Type | Quantity | Volume | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Volume | Price | Quantity & Availability | |||
10544625
|
Cytiva
28-9006-53 |
illustra Hot Start Mix RTG | 96 Reactions | 200μL |
234.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10126624
|
Cytiva
28-9006-46 |
illustra Hot Start Mix RTG | 100 Reactions | 500μL |
218.00€
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10625985
|
Cytiva
28-9006-54 |
illustra Hot Start Mix RTG | 480 Reactions | 200μL |
1039.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11349941
|
Cytiva
28-9040-91 |
illustra Hot Start Mix RTG | 8 Reactions | 200μL |
18.35€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Primer sequestration effectively blocks greater than 99% of polymerase activity during setup effectively minimizing any nonspecific priming or primer-dimer formation
- Recombinant activator protein avoids the risk of contamination from a mammalian source associated with hot start antibodies
- Quick hot start activation preserves sample integrity by avoiding extensive precycling heat treatment
- The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
Specifications
1.75 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. Ambient temperature in pouch with desiccant | |
Tube | |
illustra Hot Start Mix RTG | |
Standard PCR |